siRNA Details
Gene Symbol: Hif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010431.2 | NP_034561.2 | 6, 7 | 1202 | ||
GenBank mRNA | AF003695.1 | - | 6, 7 | 1036 | ||
AK034087.1 | - | 6, 7 | 1168 | |||
AK076395.1 | - | 4, 5 | 490 | |||
AK138944.1 | - | 6, 7 | 1069 | |||
AK150367.1 | - | 6, 7 | 1069 | |||
BC026139.1 | - | 6, 7 | 1054 | 4019056 | ||
X95580.1 | - | 6, 7 | 868 |
