siRNA Details
Gene Symbol: Hr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021877.3 | NP_068677.2 | 9, 10 | 2804 | ||
GenBank mRNA | AB221596.1 | - | 4, 5 | 489 | ||
AK076280.1 | - | 3, 4 | 289 | |||
AY312855.1 | - | 8, 9 | 2487 | |||
AY547390.1 | - | 8, 9 | 2484 | |||
AY547391.1 | - | 8, 9 | 2484 | |||
BC049182.1 | - | 9, 10 | 2804 | 6468299 | ||
Z32675.1 | - | 8, 9 | 2487 |
