siRNA Details
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010472.2 | NP_034602.1 | 4 | 763 | ||
GenBank mRNA | AK029917.1 | - | 4 | 576 | ||
AK048326.1 | - | 4 | 764 | |||
AK049400.1 | - | 4 | 750 | |||
BC031154.1 | - | 4 | 750 | 4976622 |
