siRNA Details
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010472.2 | NP_034602.1 | 7 | 1165 | ||
GenBank mRNA | AF057287.1 | - | 2 | 223 | ||
AK029917.1 | - | 7 | 978 | |||
AK048326.1 | - | 7 | 1166 | |||
AK049400.1 | - | 7 | 1152 | |||
BC031154.1 | - | 7 | 1152 | 4976622 |
