siRNA Details
Gene Symbol: Agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010472.2 | NP_034602.1 | 9, 10 | 1530 | ||
GenBank mRNA | AF057287.1 | - | 4, 5 | 588 | ||
AK029917.1 | - | 9, 10 | 1280 | |||
AK048326.1 | - | 9, 10 | 1531 | |||
AK198918.1 | - | 3, 4 | 308 | |||
BC031154.1 | - | 9, 10 | 1517 | 4976622 |
