siRNA Details
Gene Symbol: Hsd11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008289.2 | NP_032315.2 | 1, 2 | 386 | ||
GenBank mRNA | AK155153.1 | - | 1, 2 | 388 | ||
BB618868.1 | - | 1, 2 | 389 | |||
BC014753.1 | - | 1, 2 | 377 | 4235545 | ||
BC066209.1 | - | 1, 2 | 375 | 6431285 |
