siRNA Details
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010511.2 | NP_034641.1 | 2 | 259 | ||
GenBank mRNA | AF128216.1 | - | 2 | 210 | ||
AF128217.1 | - | 2 | 210 | |||
AK087525.1 | - | 2 | 248 | |||
AK150503.1 | - | 2 | 208 | |||
AK151079.1 | - | 2 | 213 | |||
AK152085.1 | - | 2 | 208 | |||
AK152384.1 | - | 2 | 208 | |||
AK153084.1 | - | 2 | 208 | |||
AK153347.1 | - | 2 | 211 | |||
AK159359.1 | - | 2 | 213 | |||
AK159660.1 | - | 2 | 153 | |||
AK159839.1 | - | 2 | 153 | |||
AK169593.1 | - | 2 | 228 | |||
AY157836.1 | - | 2 | 210 | |||
AY157837.1 | - | 2 | 210 | |||
BC132599.1 | - | 2 | 160 | 40130876 | ||
BC138617.1 | - | 2 | 160 | 8861637 | ||
BY309097.1 | - | 2 | 260 | |||
J05265.1 | - | 2 | 236 | |||
M25764.1 | - | 2 | 161 | |||
M26711.1 | - | 2 | 236 | |||
M28233.1 | - | 2 | 232 | |||
M28995.1 | - | 2 | 137 |
