siRNA Details
Gene Symbol: Inpp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF040094.2 | - | 5, 6 | 642 | ||
AK004601.1 | - | 6, 7 | 649 | |||
AK004722.2 | - | 5, 6 | 544 | |||
AY007563.1 | - | 11, 12 | 1374 | |||
BC028864.1 | - | 11, 12 | 1360 | 4457437 | ||
BU610644.1 | - | 5, 6 | 525 | |||
BU921160.1 | - | 2, 3 | 139 |
