siRNA Details
Gene Symbol: Itga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010577.3 | NP_034707.3 | 3, 4 | 696 | ||
GenBank mRNA | AK142241.1 | - | 3, 4 | 539 | ||
AK144921.1 | - | 3, 4 | 482 | |||
AK148138.1 | - | 3, 4 | 537 | |||
AK152594.1 | - | 3, 4 | 494 | |||
AK191912.1 | - | 1, 2 | 111 | |||
AK204695.1 | - | 1, 2 | 105 | |||
BC050943.1 | - | 3, 4 | 712 | 6821577 | ||
BC053431.1 | - | 4, 5 | 757 | |||
BC059829.1 | - | 3, 4 | 712 | 6821577 | ||
X79003.1 | - | 3, 4 | 653 |
