siRNA Details
Gene Symbol: Kcnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019697.3 | NP_062671.1 | 1 | 431 | ||
GenBank mRNA | AB045326.1 | - | 1037 | |||
AB093280.1 | - | 1 | 912 | |||
AF107780.1 | - | 1 | 241 | |||
AK032268.1 | - | 1222 | ||||
BC079667.1 | - | 1 | 431 | 30356567 |
