siRNA Details
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159850.1 | NP_001153322.1 | 7 | 1392 | ||
NM_001281847.1 | NP_001268776.1 | 7 | 1283 | |||
NM_001281848.1 | NP_001268777.1 | 6 | 1124 | |||
NM_010607.3 | NP_034737.2 | 7 | 1488 | |||
GenBank mRNA | AK138427.1 | - | 7 | 1164 | ||
AK140716.1 | - | 7 | 1163 | |||
AK164197.1 | - | 6 | 1124 | |||
AY736359.1 | - | 7 | 1127 | |||
BC062094.1 | - | 7 | 1106 | 6848897 | ||
CN528767.1 | - | 4 | 509 | |||
U73488.2 | - | 8 | 1565 |
