siRNA Details
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159850.1 | NP_001153322.1 | 3 | 726 | ||
NM_001281847.1 | NP_001268776.1 | 3 | 617 | |||
NM_010607.3 | NP_034737.2 | 3 | 822 | |||
GenBank mRNA | AK138427.1 | - | 3 | 498 | ||
AK140716.1 | - | 3 | 497 | |||
AY736359.1 | - | 3 | 461 | |||
BC062094.1 | - | 3 | 440 | 6848897 | ||
BY267214.1 | - | 2 | 261 | |||
U73488.2 | - | 4 | 899 |
