siRNA Details
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122733.1 | NP_001116205.1 | 16 | 2363 | ||
NM_021099.3 | NP_066922.2 | 16 | 2347 | |||
GenBank mRNA | AK046795.1 | - | 16 | 2355 | ||
AK047010.1 | - | 16 | 2360 | |||
AK145462.1 | - | 13 | 1588 | |||
AY536430.1 | - | 16 | 2310 | |||
AY536431.1 | - | 16 | 2310 | |||
BC026713.1 | - | 16 | 2342 | |||
BC052457.1 | - | 16 | 2322 | 6830637 | ||
BC075716.1 | - | 16 | 2348 | 3673641 | ||
Y00864.1 | - | 16 | 2314 |
