siRNA Details
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122733.1 | NP_001116205.1 | 13 | 2018 | ||
NM_021099.3 | NP_066922.2 | 13 | 2002 | |||
GenBank mRNA | AK046795.1 | - | 13 | 2010 | ||
AK047010.1 | - | 13 | 2015 | |||
AK145462.1 | - | 10 | 1243 | |||
AY536430.1 | - | 13 | 1965 | |||
AY536431.1 | - | 13 | 1965 | |||
BC026713.1 | - | 13 | 1997 | |||
BC052457.1 | - | 13 | 1977 | 6830637 | ||
BC075716.1 | - | 13 | 2003 | 3673641 | ||
Y00864.1 | - | 13 | 1969 |
