siRNA Details
Gene Symbol: Kit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122733.1 | NP_001116205.1 | 10 | 1694 | ||
NM_021099.3 | NP_066922.2 | 10 | 1678 | |||
GenBank mRNA | AK046795.1 | - | 10 | 1686 | ||
AK047010.1 | - | 10 | 1690 | |||
AK145462.1 | - | 7 | 919 | |||
AY536430.1 | - | 10 | 1641 | |||
AY536431.1 | - | 10 | 1641 | |||
BC026713.1 | - | 10 | 1673 | |||
BC052457.1 | - | 10 | 1653 | 6830637 | ||
BC075716.1 | - | 10 | 1679 | 3673641 | ||
Y00864.1 | - | 10 | 1645 |
