siRNA Details
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 2, 3 | 282 | ||
GenBank mRNA | AK028259.1 | - | 2, 3 | 283 | ||
AK028307.1 | - | 2, 3 | 301 | |||
AK154800.1 | - | 2, 3 | 300 | |||
AK169311.1 | - | 2, 3 | 305 | |||
AK169348.1 | - | 2, 3 | 305 | |||
AK169464.1 | - | 2, 3 | 321 | |||
AK195236.1 | - | 1, 2 | 26 | |||
AK218219.1 | - | 1, 2 | 42 | |||
BC006771.1 | - | 2, 3 | 267 | 3588534 | ||
U34228.1 | - | 1, 2 | 131 |
