siRNA Details
Gene Symbol: Kpna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008467.4 | NP_032493.1 | 17 | 1820 | ||
GenBank mRNA | AF020771.1 | - | 17 | 1528 | ||
AK147920.1 | - | 17 | 1829 | |||
BC026821.1 | - | 17 | 1615 | 3672708 | ||
BC052162.1 | - | 17 | 1821 | 6510795 | ||
CT010281.1 | - | 17 | 1528 |
