siRNA Details
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008486.2 | NP_032512.2 | 12 | 1982 | ||
GenBank mRNA | AK143746.1 | - | 12 | 1925 | ||
AK171427.1 | - | 13 | 2344 | |||
BC005431.1 | - | 12 | 1948 | 3593307 | ||
BC017011.1 | - | 12 | 2006 | 3590117 | ||
BC040792.1 | - | 12 | 2006 | 3590117 | ||
U77083.1 | - | 12 | 1914 |
