siRNA Details
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111311.1 | NP_001104781.1 | 5, 6 | 707 | ||
NM_001111312.1 | NP_001104782.1 | 6, 7 | 666 | |||
NM_008515.4 | NP_032541.1 | 14, 15 | 1223 | |||
GenBank mRNA | AF045573.1 | - | 14, 15 | 1160 | ||
AK041162.1 | - | 6, 7 | 617 | |||
AK044174.1 | - | 11, 12 | 886 | |||
AK077637.1 | - | 5, 6 | 712 | |||
AK133661.1 | - | 5, 6 | 688 | |||
AK134120.1 | - | 5, 6 | 699 | |||
BC144955.1 | - | 5, 6 | 560 | |||
BC144956.1 | - | 16, 17 | 1181 | 9053504 | ||
BC145642.1 | - | 16, 17 | 1181 | 40130945 |
