siRNA Details
Gene Symbol: Ltbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010736.3 | NP_034866.1 | 4, 5 | 664 | ||
GenBank mRNA | AI119599.1 | - | 4, 5 | 664 | ||
AK146127.1 | - | 4, 5 | 647 | |||
BC138590.1 | - | 4, 5 | 527 | 8861610 | ||
BC138591.1 | - | 4, 5 | 527 | 8861611 | ||
L38423.1 | - | 4, 5 | 664 | |||
U29173.1 | - | 4, 5 | 648 |
