siRNA Details
Gene Symbol: Cntnap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130457.2 | NP_569724.2 | 19 | 3257 | ||
GenBank mRNA | AF333770.1 | - | 19 | 3222 | ||
AK053611.1 | - | 17 | 2859 | |||
AK081891.1 | - | 19 | 3250 | |||
AK142816.1 | - | 19 | 3253 | |||
AK173255.1 | - | 19 | 3232 | |||
BC034628.1 | - | 19 | 3257 | 5369648 |
