siRNA Details
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130879.2 | NP_570949.2 | 6 | 884 | ||
GenBank mRNA | AK052297.1 | - | 6 | 826 | ||
AK133251.1 | - | 7 | 901 | |||
AK166158.1 | - | 6 | 887 | |||
AK206864.1 | - | 2 | 83 | |||
AK210453.1 | - | 2 | 83 | |||
AK217440.1 | - | 2 | 83 | |||
AK219552.1 | - | 2 | 83 | |||
AK220190.1 | - | 6 | 845 | |||
BC021769.1 | - | 6 | 843 | |||
BC145295.1 | - | 7 | 875 |
