siRNA Details
Gene Symbol: Rtn4ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130892.4 | NP_570962.2 | 6 | 1238 | ||
GenBank mRNA | AF336862.1 | - | 6 | 968 | ||
AK050324.1 | - | 6 | 1236 | |||
AK085858.1 | - | 6 | 1218 | |||
AK088029.1 | - | 6 | 1239 | |||
BC024116.1 | - | 6 | 1208 | 5100503 |
