siRNA Details
Gene Symbol: Ppargc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133249.2 | NP_573512.1 | 3, 4 | 518 | ||
GenBank mRNA | AF453324.1 | - | 3, 4 | 518 | ||
AK028464.1 | - | 4, 5 | 568 | |||
AK086111.1 | - | 3, 4 | 513 | |||
BC150699.1 | - | 3, 4 | 498 | 9087610 |
