siRNA Details
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252481.1 | NP_001239410.1 | 8 | 1257 | ||
NM_010754.5 | NP_034884.2 | 8 | 1146 | |||
GenBank mRNA | AF005743.1 | - | 7 | 874 | ||
AK007817.1 | - | 8 | 923 | |||
AY334552.1 | - | 6 | 725 | |||
BC021342.1 | - | 8 | 945 | 5066237 | ||
BC089184.1 | - | 8 | 1257 | 5322611 | ||
U60530.1 | - | 7 | 874 |
