siRNA Details
Gene Symbol: Il17rd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134437.3 | NP_602319.1 | 12 | 2088 | ||
GenBank mRNA | AF424804.1 | - | 8 | 1517 | ||
AF459444.1 | - | 12 | 2007 | |||
AF494209.1 | - | 8 | 1797 | |||
AF494210.1 | - | 12 | 2148 | |||
AK133233.1 | - | 14 | 2251 | |||
AK139904.1 | - | 12 | 2015 | |||
BC138629.1 | - | 12 | 2043 | 8861649 |
