siRNA Details
Gene Symbol: Mapkapk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010765.2 | NP_034895.1 | 4, 5 | 982 | ||
GenBank mRNA | AF039840.1 | - | 4, 5 | 996 | ||
AY533679.1 | - | 4, 5 | 290 | |||
AY533680.2 | - | 4, 5 | 290 | |||
BC019184.1 | - | 4, 5 | 404 | 5010305 |
