siRNA Details
Gene Symbol: Mcf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159485.1 | NP_001152957.1 | 26 | 3094 | ||
NM_001159486.1 | NP_001152958.1 | 26 | 3094 | |||
NM_178076.3 | NP_835177.2 | 26 | 2997 | |||
GenBank mRNA | AK122259.1 | - | 17 | 1902 | ||
AK147252.1 | - | 23 | 3201 | |||
AK170186.1 | - | 26 | 3055 | |||
AK170558.1 | - | 26 | 3028 | |||
BC029315.1 | - | 7 | 680 | |||
BC058622.1 | - | 26 | 2932 | 5704822 | ||
BQ555281.1 | - | 1 | 77 | |||
CF618259.1 | - | 3 | 213 | |||
DQ978380.1 | - | 25 | 2848 | |||
S76838.1 | - | 27 | 3291 |
