siRNA Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170537.1 | NP_001164008.1 | 6 | 1041 | ||
NM_025282.3 | NP_079558.1 | 6 | 1035 | |||
GenBank mRNA | AK009139.1 | - | 6 | 875 | ||
AK047994.1 | - | 4 | 620 | |||
AK077603.1 | - | 5 | 831 | |||
AK132573.1 | - | 6 | 870 | |||
BC026841.1 | - | 5 | 773 | 4481227 | ||
BC037731.1 | - | 5 | 763 | 4500786 | ||
BC057650.1 | - | 5 | 752 | 4500786 | ||
L13171.1 | - | 4 | 602 |
