siRNA Details
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 13 | 2148 | ||
NM_010789.3 | NP_034919.1 | 12 | 2053 | |||
GenBank mRNA | AB221660.1 | - | 11 | 1316 | ||
AK134321.1 | - | 13 | 2143 | |||
AK140748.1 | - | 13 | 2152 | |||
AK146680.1 | - | 9 | 956 | |||
AK168684.1 | - | 13 | 2141 | |||
BC034136.1 | - | 1 | 1238 | |||
U33629.1 | - | 13 | 1859 | |||
U33630.1 | - | 12 | 1764 |
