siRNA Details
Gene Symbol: Mlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026810.2 | NP_081086.2 | 8 | 729 | ||
GenBank mRNA | AF250844.1 | - | 8 | 645 | ||
AK038265.1 | - | 8 | 722 | |||
AK145646.1 | - | 8 | 676 | |||
AK163861.1 | - | 8 | 664 | |||
AK168849.1 | - | 8 | 668 | |||
BC021815.1 | - | 8 | 683 | 4020489 |
