siRNA Details
Gene Symbol: Mmp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008607.2 | NP_032633.1 | 4 | 582 | ||
GenBank mRNA | AK150728.1 | - | 4 | 569 | ||
AK151626.1 | - | 4 | 569 | |||
AK152507.1 | - | 4 | 568 | |||
AK152916.1 | - | 4 | 569 | |||
AK153458.1 | - | 4 | 567 | |||
AK155188.1 | - | 4 | 568 | |||
AK159921.1 | - | 4 | 569 | |||
AK160819.1 | - | 4 | 567 | |||
BC125320.1 | - | 4 | 582 | 40129835 | ||
BC125322.1 | - | 4 | 582 | 40129837 | ||
X66473.1 | - | 4 | 551 |
