siRNA Details
Gene Symbol: Msh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010830.2 | NP_034960.1 | 4 | 2366 | ||
GenBank mRNA | AK088259.1 | - | 4 | 2355 | ||
BC035274.1 | - | 2 | 1764 | |||
BC051160.1 | - | 4 | 2311 | 4953238 | ||
BC051634.1 | - | 4 | 2321 | 6518577 |
