siRNA Details
Gene Symbol: Msh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010830.2 | NP_034960.1 | 4 | 2982 | ||
GenBank mRNA | AK010643.1 | - | 1 | 143 | ||
AK088259.1 | - | 4 | 2971 | |||
BC035274.1 | - | 2 | 2380 | |||
BC051160.1 | - | 4 | 2927 | 4953238 | ||
BC051634.1 | - | 4 | 2937 | 6518577 | ||
U42190.1 | - | 4 | 2958 |
