siRNA Details
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010832.5 | NP_034962.2 | 9 | 1262 | ||
GenBank mRNA | AF117066.2 | - | 9 | 1264 | ||
AK077794.1 | - | 3 | 522 | |||
AK089368.1 | - | 9 | 1345 | |||
AK146208.1 | - | 9 | 1198 | |||
AK148898.1 | - | 9 | 1239 |
