siRNA Details
Gene Symbol: Neo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042752.1 | NP_001036217.1 | 3 | 966 | ||
NM_008684.2 | NP_032710.2 | 3 | 966 | |||
GenBank mRNA | AK084609.1 | - | 3 | 955 | ||
BC054540.1 | - | 3 | 950 | 6406007 | ||
Y09535.1 | - | 3 | 833 |
