siRNA Details
Gene Symbol: Neo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042752.1 | NP_001036217.1 | 4 | 1083 | ||
NM_008684.2 | NP_032710.2 | 4 | 1083 | |||
GenBank mRNA | AK084609.1 | - | 4 | 1072 | ||
BC054540.1 | - | 4 | 1067 | 6406007 | ||
Y09535.1 | - | 4 | 950 |
