siRNA Details
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010897.2 | NP_035027.1 | 57 | 8297 | ||
GenBank mRNA | AK033306.1 | - | 17 | 2144 | ||
AK050818.1 | - | 10 | 1044 | |||
L10370.1 | - | 56 | 8142 |
