siRNA Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010901.2 | NP_035031.2 | 9 | 3311 | ||
GenBank mRNA | AK088536.1 | - | 1 | 165 | ||
AK131919.1 | - | 7 | 1723 | |||
AK134054.1 | - | 9 | 3230 | |||
AK220187.1 | - | 9 | 3084 | |||
BC021835.1 | - | 6 | 1515 | |||
BC120900.1 | - | 9 | 3030 | 40085687 | ||
BC120901.1 | - | 9 | 3030 | 40085688 | ||
D85612.1 | - | 9 | 3027 | |||
EU887626.1 | - | 9 | 2999 | |||
EU887627.1 | - | 9 | 2999 | |||
EU887629.1 | - | 9 | 3023 | |||
EU887630.1 | - | 9 | 3023 | |||
U28807.1 | - | 9 | 2990 |
