siRNA Details
Gene Symbol: Nfkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008689.2 | NP_032715.2 | 10 | 1265 | ||
GenBank mRNA | AB119195.1 | - | 8 | 763 | ||
AB221614.1 | - | 8 | 770 | |||
AK052726.1 | - | 5 | 573 | |||
AY521462.1 | - | 8 | 763 | |||
AY521463.1 | - | 8 | 763 | |||
BC050841.1 | - | 10 | 1265 | |||
BC138535.1 | - | 9 | 842 | 8861555 | ||
BC138536.1 | - | 9 | 842 | 8861556 | ||
BC145317.1 | - | 10 | 859 | |||
M57999.1 | - | 10 | 1054 |
