siRNA Details
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081453.1 | NP_001074922.1 | 16 | 3375 | ||
NM_001286079.2 | NP_001273008.1 | 17 | 3557 | |||
NM_001286080.2 | NP_001273009.1 | 18 | 3604 | |||
NM_008697.4 | NP_032723.2 | 18 | 3604 | |||
GenBank mRNA | AK129394.1 | - | 18 | 3494 | ||
AK147187.1 | - | 1 | 360 | |||
AY515727.1 | - | 16 | 3354 | |||
BC137789.1 | - | 17 | 3442 | 8860808 | ||
BC144913.1 | - | 18 | 3489 | 9053461 | ||
BC144914.1 | - | 18 | 3489 | |||
U40342.1 | - | 18 | 3517 |
