siRNA Details
Gene Symbol: Nos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008713.4 | NP_032739.3 | 3 | 409 | ||
GenBank mRNA | AK077896.1 | - | 3 | 411 | ||
AK143221.1 | - | 2 | 181 | |||
AK158106.1 | - | 3 | 422 | |||
BC052636.1 | - | 3 | 396 | 6530765 | ||
U53142.1 | - | 3 | 397 |
