siRNA Details
Gene Symbol: Notch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008716.2 | NP_032742.1 | 12 | 1955 | ||
GenBank mRNA | X74760.1 | - | 12 | 1955 |

More Information
Other Species Targeted

Gene Ontology Categories:
