siRNA Details
Gene Symbol: Uhrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111078.1 | NP_001104548.1 | 5 | 845 | ||
NM_001111079.1 | NP_001104549.1 | 5 | 989 | |||
NM_001111080.1 | NP_001104550.1 | 5 | 845 | |||
NM_010931.3 | NP_035061.3 | 5 | 989 | |||
GenBank mRNA | AF274046.1 | - | 5 | 902 | ||
AK143688.1 | - | 5 | 814 | |||
AK145376.1 | - | 5 | 811 | |||
AK145419.1 | - | 5 | 811 | |||
AK145543.1 | - | 5 | 929 | |||
AK145583.1 | - | 5 | 811 | |||
AK146951.1 | - | 5 | 962 | |||
AK147046.1 | - | 5 | 929 | |||
AK150489.1 | - | 5 | 845 | |||
AK151701.1 | - | 5 | 811 | |||
AK151837.1 | - | 5 | 845 | |||
AK152930.1 | - | 5 | 845 | |||
AK153083.1 | - | 5 | 845 | |||
BC022167.1 | - | 5 | 917 | 4017286 | ||
D87908.1 | - | 5 | 900 |
