siRNA Details
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008720.2 | NP_032746.2 | 14 | 2355 | ||
GenBank mRNA | AF003348.1 | - | 14 | 2332 | ||
AK154282.1 | - | 14 | 2275 | |||
AK155379.1 | - | 14 | 2275 | |||
BC052437.1 | - | 14 | 2355 | 6405214 | ||
BC054539.1 | - | 14 | 2355 | 6405214 |
