siRNA Details
Gene Symbol: Nrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008737.2 | NP_032763.2 | 9 | 1815 | ||
GenBank mRNA | AK082992.1 | - | 9 | 1868 | ||
AK141964.1 | - | 9 | 1607 | |||
AK148372.1 | - | 9 | 1791 | |||
AK149752.1 | - | 4 | 396 | |||
AK159219.1 | - | 9 | 1606 | |||
BC060129.1 | - | 9 | 1595 | 6409596 | ||
D50086.1 | - | 9 | 1675 |
