siRNA Details
Gene Symbol: Nrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008737.2 | NP_032763.2 | 3 | 840 | ||
GenBank mRNA | AK082992.1 | - | 3 | 893 | ||
AK135428.1 | - | 3 | 517 | |||
AK141964.1 | - | 3 | 632 | |||
AK148372.1 | - | 3 | 816 | |||
AK159219.1 | - | 3 | 631 | |||
AK164378.1 | - | 3 | 842 | |||
BC060129.1 | - | 3 | 620 | 6409596 | ||
D50086.1 | - | 3 | 700 |
