siRNA Details
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037127.2 | NP_001032204.2 | 15, 16 | 2139 | ||
NM_001037128.1 | NP_001032205.1 | 15, 16 | 2094 | |||
NM_001037129.1 | NP_001032206.1 | 14, 15 | 2070 | |||
NM_001037130.1 | NP_001032207.1 | 13, 14 | 2040 | |||
NM_010944.2 | NP_035074.2 | 14, 15 | 2064 | |||
GenBank mRNA | AB691772.1 | - | 13, 14 | 1890 | ||
BC100342.1 | - | 14, 15 | 1994 | 30935290 | ||
X86444.1 | - | 13, 14 | 1968 |
