siRNA Details
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037127.2 | NP_001032204.2 | 16 | 2772 | ||
NM_001037128.1 | NP_001032205.1 | 16 | 2727 | |||
NM_001037129.1 | NP_001032206.1 | 15 | 2703 | |||
NM_001037130.1 | NP_001032207.1 | 14 | 2673 | |||
NM_010944.2 | NP_035074.2 | 15 | 2697 | |||
GenBank mRNA | AB691772.1 | - | 14 | 2523 | ||
AY955296.1 | - | 16 | 2622 | |||
AY955297.1 | - | 16 | 2577 | |||
AY955298.1 | - | 15 | 2553 | |||
AY956403.1 | - | 15 | 2547 | |||
AY956404.1 | - | 14 | 2523 | |||
BC100342.1 | - | 15 | 2627 | 30935290 | ||
U37708.1 | - | 14 | 2671 | |||
U37709.1 | - | 15 | 2695 | |||
X86444.1 | - | 14 | 2601 |
