siRNA Details
Gene Symbol: Bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019788.3 | NP_062762.1 | 3 | 415 | ||
GenBank mRNA | AK038381.1 | - | 3 | 393 | ||
AK077720.1 | - | 3 | 397 | |||
AK147116.1 | - | 3 | 418 | |||
AK160580.1 | - | 3 | 385 | |||
AK170964.1 | - | 3 | 346 | |||
AK172571.1 | - | 3 | 396 | |||
BC016554.1 | - | 3 | 394 | |||
BC085260.1 | - | 3 | 384 | 6437935 |
